Would you like to log in again?
Anti-Caspase 3 Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP096-05-RUO RP096-RUO RP096-05 RP096-01 RP096-01-RUO RP096
76635-112EA 838.4 USD InStock
CA76635-110 76635-112 76635-110 CA76635-112 CA76635-108 76635-108
Anti-Caspase 3 Rabbit Polyclonal Antibody
Antibodies
This antibody is specific to a 32 kDa protein, which is identified as Caspase 3. Caspase 3 is associated with induction of apoptosis. Caspase 3 is ubiquitously expressed and is synthesized as an inactive proenzyme.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty