Would you like to log in again?
Anti-Factor XIIIa Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP103 RP103-01 RP103-RUO RP103-05 RP103-01-RUO RP103-05-RUO
76635-120EA 105.1 USD InStock
76635-120 76635-124 CA76635-122 76635-122 CA76635-120 CA76635-124
Anti-Factor XIIIa Rabbit Polyclonal Antibody
Antibodies
This antibody is specific to 160 kDa protein known as Factor XIII-A. Factor XIII is a β-globulin found in plasma and is composed of two subunits, Factor XIII-A and Factor XIII-B. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is a dermal dendrocyte marker and v variable reaction with these types of tumors.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty