Would you like to log in again?
Anti-Granzyme B Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP105-RUO RP105-05-RUO RP105 RP105-01-RUO RP105-01 RP105-05
76635-130EA 855.12 USD InStock
76635-130 CA76635-130 CA76635-126 76635-128 76635-126 CA76635-128
Anti-Granzyme B Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 32 kDa protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTL) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned. Human granzyme B is involved in target cell apoptosis during lymphocyte-mediated cytotoxicity. This antibody is useful in the localization of granzyme B containing lytic granules and for the characterization of activated CTL or NK cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty