Would you like to log in again?
Anti-Amyloid Precursor Protein (APP) Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP123-01 RP123-RUO RP123-05 RP123-05-RUO RP123-01-RUO RP123
76635-142EA 501.6 USD InStock
CA76635-140 76635-142 76635-140 CA76635-142 76635-138 CA76635-138
Anti-Amyloid Precursor Protein (APP) Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 95 to 100 kDa protein. This antibody recognizes amyloid precursor proteins APP695, APP751 and APP770. Amyloid precursor protein and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty