Would you like to log in again?
Anti-Claudin 1 Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP153 RP153-05-RUO RP153-01-RUO RP153-05 RP153-RUO RP153-01
76635-156EA 117.04 USD InStock
CA76635-160 76635-156 CA76635-158 CA76635-156 76635-158 76635-160
Anti-Claudin 1 Rabbit Polyclonal Antibody
Antibodies
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact, made up of a network of strands to act as a molecular gasket for preventing the leakage of ions, water etc., between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different claudin composition.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty