Would you like to log in again?
Anti-P40 Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP163-RUO RP163-01-RUO RP163-05-RUO RP163 RP163-01 RP163-05
76635-164EA 459.8 USD InStock
CA76635-162 76635-164 76635-162 CA76635-166 CA76635-164 76635-166
Anti-P40 Rabbit Polyclonal Antibody
Antibodies
p40 is a relatively unknown antibody that recognizes ΔNp63-a p63 isoform suggested to be highly specific for squamous/basal cells. In a recent study, p40 is equivalent to p63 in sensitivity for squamous cell carcinoma, but it is markedly superior to p63 in specificity1, which eliminates a potential pitfall of misinterpreting a p63-positive adenocarcinoma or unsuspected lymphoma as squamous cell carcinoma. These findings strongly support the routine use of p40 in place of p63 for the diagnosis of pulmonary squamous cell carcinoma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty