Would you like to log in again?
Anti-Smad4 Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP173-01-RUO RP173-RUO RP173-05 RP173-01 RP173 RP173-05-RUO
76635-186EA 82.41 USD InStock
76635-186 CA76635-188 CA76635-186 76635-188 CA76635-190 76635-190
Anti-Smad4 Rabbit Polyclonal Antibody
Antibodies
SMADs are members of the MAD-related family of molecules. MAD-related proteins are a family of intracellular proteins that are essential components in the signaling pathways of the serine/threonine kinase receptors of the transforming growth factor beta superfamily. Alterations in the SMAD4 gene was primarily discovered in pancreatic cancer (duct adenocarcinoma) but occur in a variety of cancers such as colorectal cancer, gastric cancer, prostate cancer, melanomas, head and neck cancers and many others, though with higher frequencies in gastrointestinal tract cancers. Loss of SMAD4 expression in tumors has also been shown to affect cancer progression and therapy, such as reduced response to adjuvant chemotherapy.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty