Would you like to log in again?
Anti-Caspase 1 Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP175-01 RP175-RUO RP175-05 RP175-01-RUO RP175-05-RUO RP175
76635-196EA 894.52 USD InStock
CA76635-194 76635-196 76635-194 CA76635-192 CA76635-196 76635-192
Anti-Caspase 1 Rabbit Polyclonal Antibody
Antibodies
Caspase 1 and is expressed as a proenzyme in many tissues. Active Caspase 1 is a tetramer of two subunits p10 and p20 in 2:2 ratio. Overexpression of caspase 1 can induce apoptosis in fibroblast, which can be inhibited by overexpression of Crm A, a protein from pox virus.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty