Would you like to log in again?
Anti-Uroplakin II (UPK2) Rabbit Antibody
Supplier: Diagnostic BioSystems
RP177-05 RP177 RP177-RUO RP177-01-RUO RP177-01 RP177-05-RUO
76635-204EA 204.23 USD InStock
CA76635-208 76635-204 76635-208 CA76635-206 76635-206 CA76635-204
Anti-Uroplakin II (UPK2) Rabbit Antibody
Antibodies
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. Uroplakins are markers of terminally differentiated urothelium. Uroplakin II (UPII) is a newly described sensitive marker for urothelial carcinoma (UC). The expression profile of UPII in different types of UC and its utility in the diagnostic setting are needed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty