Would you like to log in again?
Anti-Uroplakin ll/lll cocktail Multiple Recombinant Antibody
Supplier: Diagnostic BioSystems
RPM001 RPM001-05 RPM001-RUO RPM001-01-RUO RPM001-01 RPM001-05-RUO
76635-212EA 539.82 USD InStock
76635-212 CA76635-210 76635-210 CA76635-214 76635-214 CA76635-212
Anti-Uroplakin ll/lll cocktail Multiple Recombinant Antibody
Antibodies
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. In non-neoplastic mammalian urothelium, UPs are expressed in the luminal surface plasmalemma of superficial (umbrella) cells, Uroplakin II/III cocktail is specific for tumors of urothelial origin and, when used in combination with other markers, can aid in the diagnosis of primary and metastatic tumors.
  • Hosts: Mouse and Rabbit
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty