Would you like to log in again?
SITVue /DAB Detection System, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
SIT-100D SIT-100D-RUO
76635-216EA 674.77 USD InStock
76635-216 CA76635-216
SITVue /DAB Detection System, Diagnostic BioSystems
Protein Linking and Labeling Systems
SITVue /DAB detection system is a powerful Intensification system that significantly enhances chromogenic signals. This system can be integrated into standard immunohistochemistry (IHC) staining methods.
SITVue /DAB Detection System is an enzyme-mediated reaction that utilizes horseradish peroxidase (HRP) to catalyze the deposition of two separate Linkers applied sequentially onto tissue sections or cell preparation. The deposited Linkers can be detected with a streptavidin-peroxidase conjugate followed by a reaction with a peroxidase substrate/chromogen solution such as diaminobenzidine (DAB).
SITVue /DAB Detection System results in a significant increase in sensitivity compared to standard IHC detection methods, while maintaining similar specificity.
Delivery information: Kit contains: Tissue primer (10 ml), Linker 1 (10 ml), Linker 2 (10 ml), Tracer (10 ml), Stable DAB/Plus buffer (10 ml), Stable DAB/Plus chromogen (15 ml), Empty mixing bottle for Stable DAB/Plus (1 ml) and 15 ml bottle.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty