Would you like to log in again?
Bioquell BQ-EMS Bio-Decontamination System
Supplier: Bioquell
VWRBQEMS
76635-460EA 106499.99 USD InStock
76635-460
Bioquell BQ-EMS Bio-Decontamination System
Disinfection Systems
The Bioquell BQ-EMS is a mobile bio-decontamination system. It uses Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703) to kill microorganisms on exposed surfaces within emergency response vehicles by creating a 6-log sporicidal kill. Its setup allows for rapid transport and maneuverability.
  • Mobile, lightweight and versatile bio-decontamination system
  • 6-log sporicidal kill using Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number: 72372-1-86703)
  • Automated cycles after entering in dimensions
  • RFID bottle reading for safety and quality control
  • No wipe downs or additional steps after cycle completion
  • Requires only a single cycle
  • Cycles in as fast as 45 minutes
The Bioquell BQ-EMS is a mobile bio-decontamination system . The Bioquell technology, when utilized with Bioquell Hydrogen Peroxide Sterilant, kills microorganisms on exposed surfaces within emergency response vehicles, achieving a 6-log sporicidal kill. Its setup allows for rapid transport and maneuverability.
Whether at the end of a shift or during routine maintenance, hydrogen peroxide vapor technology from Ecolab vaporizes Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703) to kill microorganisms on exposed surfaces. All that is required to remove during a bio-decontamination cycle are absorbent materials such as linens, and items sensitive to hydrogen peroxide such as glucometers. Otherwise, all drawers and cabinets should remain open to expose as much surface area as possible. After a single cycle, the vehicle is immediately ready for use as there are no additional steps in order to reclaim the space.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty