Would you like to log in again?
Bioquell L-4, Hydrogen Peroxide Vapor Bio-Decontamination Generator
Supplier: Bioquell
VWRBQL4
76635-462EA 106499.99 USD InStock
76635-462
Bioquell L-4, Hydrogen Peroxide Vapor Bio-Decontamination Generator
Disinfection Systems
The Bioquell L-4 is a mobile Hydrogen Peroxide Vapor bio-decontamination generator that can be connected to enclosures, equipment, isolators and more in your facility or, with additional accessories, be used for room/zone bio-decontamination.
  • Create validated cycles for 6-log sporicidal kill on exposed surfaces utilizing Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703)
  • Adaptable to various equipment types including Biosafety cabinets, closed systems and isolators, cage and rack washers, pass-throughs, material airlocks, freeze dryers and more
  • Optional room bio-decontamination through distribution head for areas smaller than 350m3 (12360 ft3)
  • Utilizes EPA registered sterilant 35% w/w aqueous hydrogen peroxide solution
  • Designed with RFID label technology for traceability and auditing
  • 21 CFR Part 11 audit trail compliance package
  • Integration testing for BMS and more complex systems
  • Cycle recording (thermal printer and solutions for real time cycle data, including electronic records in pdf format)
  • Remote start stop integration and Automation
  • Supports GMP compliance
The Bioquell L-4 is a mobile Hydrogen Peroxide Vapor generator that can be connected to enclosures, equipment, isolators and more in your facility or, with additional accessories, be used for room/zone bio-decontamination.
Disinfect equipment or small enclosure systems, open spaces or rooms, and more using the multipurpose Bioquell L-4. Bio-decontamination can be achieved through quick and direct connections to equipment and ports, open spaces, or for custom engineering solutions. Bio-decontaminate isolators, BSCs, lyophilizers, pass-throughs, rack washers and more when connected to the Bioquell L-4.
Room bio-decontamination options further showcase the system’s versatility. Use the Bioquell L-4 inside a room by attaching a distribution head, or outside of a zone with connection solutions to meet your needs.
Ecolab’s Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number: 72372-1-86703) creates a 6-log sporicidal kill on exposed surfaces when used with Ecolab's Bioquell bio-decontamination equipment. You can operate Bioquell systems at ambient room temperature and relative humidity, without the need to attain significantly reduced humidity commonly associated with other hydrogen peroxide bio-decontamination systems.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty