Would you like to log in again?
InTemp® CX402-T Temperature Data Loggers with Probe and Glycol Bottle
Supplier: Onset
InTemp®
CX402-T430 CX402-T415 CX402-T205 CX402-T215 CX402-T230 CX402-T405
76635-552EA 191.48 USD InStock
76635-552 76635-546 76635-556 76635-544 76635-554 76635-548
InTemp® CX402-T Temperature Data Loggers with Probe and Glycol Bottle
Data Loggers
InTemp CX402-Txxx (with glycol bottle) data loggers monitor temperatures in refrigerators, freezers and other temperature-controlled environments to comply with global GDP requirements.
  • Integrates with mobile and cloud platforms for a 21 CFR part 11 compliant solution
  • Easy-to-read display with current and daily min/max temperatures
  • Bluetooth transmission of data and alarm states
  • Logger setup and download of data in seconds – without a PC or cable
  • Real-time alerts and automatic data downloads with optional CX gateway device
  • ±0.5 °C accuracy for reliable temperature readings
  • Battery-powered to guard against local power loss
InTemp CX402-Txxx (with glycol bottle) data loggers monitor temperatures in refrigerators, freezers and other temperature-controlled environments to comply with global GDP requirements. The loggers communicate wirelessly via bluetooth to mobile devices. Using the InTemp app, you can easily view data, check logger status, set alarms, and create and share secure PDF reports for streamlined reporting and regulatory compliance. All loggers feature a built-in LCD display to view current and daily min and max temperatures, advanced audible and visual alarm capabilities for notification of temperature excursions, an internal sensor for ambient temperature monitoring, and a single-point NIST certification of calibration for both the logger body and probe.
With the optional CX gateway, you can get remote notifications of temperature excursions and automatically download data from loggers within range of the device. Learn more about the CX gateway.
Models are available with a 2 or 4 meter external temperature probe attached to a glycol bottle used to insulate the probe and delay thermal response times.
Certifications: Calibration includes a single point NIST certificate, probe and internal ambient sensor from ISO17025/ANAB certified lab valid for one year. GDP/GMP compliant. DO160 compliant. Follows GDP/GMP, FDA and/or EU/CE guidelines.
Accessories information: The CX gateway (76635-574; sold separately) links your InTemp data loggers seamlessly to the InTempConnect cloud-based data platform to provide real-time alert notifications for InTemp loggers, automatically.
Delivery information: Batteries Included. NIST certificate included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty