Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
InTemp® Wireless Temperature Data Logger with Internal Sensor
Supplier: Onset
InTemp®
CX403
76635-568EA 133.52 USD InStock
76635-568
InTemp® Wireless Temperature Data Logger with Internal Sensor
Data Loggers
The InTemp CX403 data logger features an internal sensor for monitoring ambient temperature.
  • Internal sensor only (no probe)
  • Refrigerated to ambient monitoring
  • Easy-to-read display with current and daily min/max temperatures
  • Logger setup and download of data in seconds – without a PC or cable
  • ±0.5 °C accuracy for reliable temperature readings
  • GDP/GMP compliant
  • Integrates with mobile and cloud platforms for a 21 CFR part 11 compliant solution
  • Bluetooth transmission of data and alarm states
  • Real-time alerts and automatic data downloads with optional CX gateway device
  • Battery-powered to guard against local power loss
The InTemp CX403 data logger features an internal sensor for monitoring ambient temperature. Communicating wirelessly via bluetooth to mobile devices, this logger features a built-in LCD display to view ambient temperature, advanced audible and visual alarm capabilities to notify of temperature excursions, and a NIST certification of calibration.
With the optional CX gateway, you can get remote notifications of temperature excursions and automatically download data from loggers within range of the device.
Certifications: Calibration includes a single point NIST certificate of calibration for the internal amibent sensor only from ISO17025/ANAB certified lab, valid for one year. GDP/GMP compliant. DO160 compliant.
Accessories information: The CX gateway (76635-574; sold separately) links your InTemp data loggers seamlessly to the InTempConnect cloud-based data platform to provide real-time alert notifications for InTemp loggers, automatically.
Delivery information: Batteries included. NIST certificate included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty