Would you like to log in again?
InTemp® Temp/Relative Humidity Data Logger
Supplier: Onset
InTemp®
CX450
76635-572EA 164.57 USD InStock
76635-572
InTemp® Temp/Relative Humidity Data Logger
Data Loggers
The InTemp CX450 temp/RH data logger monitors temperature and relative humidity in storage and transportation environments.
  • Easy logger setup and data download in seconds
  • Integrates with mobile and cloud platforms for a 21 CFR part 11 compliant solution
  • LCD display, with ability to clear min/max temp and RH readings
  • Unique audible alarms for temp, RH, and combined temp/RH
  • AAA battery-powered to guard against local power loss
  • InTemp mobile app to configure logger, view data, and access the free InTempConnect cloud software service
  • Real-time alerts and automatic data downloads with optional InTemp CX gateway device
The CX450 uses bluetooth technology to wirelessly communicate with the InTemp CX gateway and/or the InTemp mobile app, and sends data to the InTempConnect cloud platform. The external sensor provides highly accurately temperature (−30 to +70 °C) and humidity (0 to 95%) readings. This Temp/RH logger also features a built-in LCD display to view temperature and relative humidity readings, visual and unique audible alarms, and battery level. Users can also view and display minimum and maximum readings, without affecting logged data.
By adding the InTemp CX gateway, users can get text or email notifications of temperature and/or humidity excursions, as well as automatic data downloads from loggers within range of the CX gateway device.
Certifications: Calibration includes two-point NIST certificate of calibration, from ISO17025/ANAB certified lab valid for one year. GDP/GMP/ISO9001 compliant. DO160 compliant.
Accessories information: The CX gateway (76635-574; sold separately) links your InTemp data loggers seamlessly to the InTempConnect cloud-based data platform to provide real-time alert notifications for InTemp loggers, automatically.
Delivery information: Batteries included. NIST certificate included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty