Would you like to log in again?
VWR® Macro Pipette Tips
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
4521-S1-021AV 4411-SF-521AV 4410-00-321AV 4420-00-321AV 4531-SF-621AV 4530-00-411AV 4531-S0-021AV 4421-S0-021AV 4520-01-411AV 4411-S0-021AV 4421-SF-021AV 4420-0F-521AV 4530-0F-621AV 4410-0F-521AV
76635-628CS 118.25 USD InStock
76635-628 76635-656 76635-666 76635-644 76635-642 76635-664 76635-648 76635-668 76635-674 76635-630 76635-662 76635-640 76635-672 76635-660
VWR® Macro Pipette Tips
Pipette Tips
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty