Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sterile SatPax® MicroSeal®-VP Pre-wetted Sealed Edge Wipes, 100% Polyester Knit

Supplier: Berkshire
SatPax® MicroSeal®-VP
SSPVP00124 SSPVPR00124
76635-706CS 857.75 USD InStock
76635-706 77581-280
Sterile SatPax® MicroSeal®-VP Pre-wetted Sealed Edge Wipes, 100% Polyester Knit
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Sterile SatPax® MicroSeal®-VP is an ultrasonically sealed edge cleanroom laundered wipe recommended for ISO Class 3 and above environments. This wipe is composed of 100% continuous filament polyester knit fabric.

  • Pre-wetted with 70% USP Grade IPA
  • Ultrasonically sealed edge for reduced fiber contamination
  • Laundered and packaged in Berkshire's ISO Class 4 cleanroom with lot number on each pack
  • Lightweight, high absorbency material design reduces actual cost in use compared to heavier weight products
  • Critically low particles, fibers, ions, and extractables

Sterile SatPax® MicroSeal®-VP wipes are pre-wetted with a consistent concentration of IPA and DI water and a 100% IPA option. This combination of properties provides Sterile SatPax® MicroSeal ®-VP with the highest cleanliness, abrasion resistance, and chemical compatibility required for cleaning critical environments. The pre-wetted format offers a cost-effective and easy-to-use solution versus traditional bulk handling of solvents and maintenance of squirt bottles.

Certifications: ISO Class 3 - 5, Cleanroom Class 1 - 100. EU Grade B/C/D.

Packaging: Wipes are packaged 30 sheets/pack, 24 packs/case (J-fold).

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty