Would you like to log in again?
Magid® D-ROC® DX+ Technology® DXPG53 18-Gauge VersaTek Grip™ Palm Coated Coreless Work Gloves, Cut Level A5
Supplier: Magid
Magid® D-ROC® DX+ Technology®
DXPG535 DXPG539 DXPG5310 DXPG536 DXPG5312 DXPG538 DXPG5311 DXPG537
76635-728PR 20.6 USD InStock
76635-728 76635-738 76635-732 76635-730 76635-726 76635-736 76635-724 76635-734
Magid® D-ROC® DX+ Technology® DXPG53 18-Gauge VersaTek Grip™ Palm Coated Coreless Work Gloves, Cut Level A5
Gloves
The Magid® D-ROC® DXPG53 18-gauge work glove features DX+ Technology® for high cut protection with the flexible comfort of a coreless shell and a VersaTek Grip™ palm coating for extreme durability, dexterity, and grip in any environment.
  • DX+ Technology® feels cool to the touch which keeps the wearer cool and comfortable while working
  • Intelligent VersaTek Grip™ adapts to amplify grip ability in wet, dry, or oily environments
  • Reinforced thumb saddle enhances durability at a critical wear point
  • Touchscreen compatible to maintain user safety while operating electronic devices
  • Shield graphic is color-coded by cut level to easily identify cut protection from a distance
  • Machine washable for longer service life
  • ANSI Level A5 cut resistance, ANSI Level 6 abrasion resistance, and ANSI Level 3 puncture resistance
VersaTek is a revolutionary coating technology engineered for maximum comfort and flexibility with stabilizers that enhance performance and durability. VersaTek Adaptive Coating Technology delivers unmatched durability while flexing with natural hand movements for precision handling of small parts. The DXPG53 achieves an A5 cut level, is cool to the touch, and is 30% lighter in weight than comparable cut-resistant materials, all with a coating that reads the environment and adapts to what is needed to get the job done.
Coreless DX+ Technology® uses non-irritating strength enhancing micro-particles to achieve higher levels of cut protection without the discomfort that fiberglass or steel can cause. Premium 18-gauge DX+ Technology® engineered yarn delivers extreme comfort that is 30% lighter than a traditional HPPE of the same cut level.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty