Would you like to log in again?
Magid® D-ROC® DX+ Technology® DXPG59Q 18-Gauge Double-Dipped Foam Nitrile 3/4 Coated Coreless Work Gloves, Cut Level A5
Supplier: Magid
Magid® D-ROC® DX+ Technology®
DXPG59Q10 DXPG59Q7 DXPG59Q12 DXPG59Q11 DXPG59Q9 DXPG59Q8
76635-762PR 22.8 USD InStock
76635-762 76635-760 76635-754 76635-752 76635-758 76635-756
Magid® D-ROC® DX+ Technology® DXPG59Q 18-Gauge Double-Dipped Foam Nitrile 3/4 Coated Coreless Work Gloves, Cut Level A5
Gloves
The Magid® D-ROC® DXPG59Q 18-gauge double-dipped foam nitrile 3/4 coated work gloves features DX+ Technology®, pairing high cut protection with the flexible comfort of a coreless shell.
  • DX+ Technology® feels cool to the touch which keeps the wearer cool and comfortable while working
  • Foam nitrile palm coating channels oil away for superb grip in light oil applications
  • 3/4 Nitrile undercoat protects against liquids, oils, and solvents
  • Shield graphic is color-coded by cut level to easily identify cut protection from a distance
  • Machine washable for longer service life
  • ANSI Level A5 cut resistance, ANSI Level 5 abrasion resistance, and ANSI Level 4 puncture resistance
These non-irritating work gloves made of proprietary yarn achieve higher levels of cut protection without irritation. The DXPG59Q pairs level A5 cut protection with a 3/4 nitrile undercoat and foam nitrile palm coating to protect against liquids, oils, and solvents while maintaining a strong grip.
Coreless DX+ Technology® uses non-irritating strength enhancing micro-particles to achieve higher levels of cut protection without the discomfort that fiberglass or steel can cause. The premium 18-gauge DX+ Technology® engineered yarn delivers extreme comfort that is 30% lighter than a traditional HPPE of the same cut level.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty