Would you like to log in again?
Magid® D-ROC® DX+ Technology® DXPG62 Polyurethane Palm Coated Coreless Work Gloves, 13 Gauge, Cut Level A6
Supplier: Magid
Magid® D-ROC® DX+ Technology®
DXPG625 DXPG6210 DXPG626 DXPG6212 DXPG628 DXPG629 DXPG6211 DXPG627
76635-772PR 20.6 USD InStock
76635-772 76635-770 76635-766 76635-776 76635-764 76635-774 76635-768 76635-778
Magid® D-ROC® DX+ Technology® DXPG62 Polyurethane Palm Coated Coreless Work Gloves, 13 Gauge, Cut Level A6
Gloves
The Magid® D-ROC® DXPG62 lightweight 13-gauge cut-resistant work gloves features DX+ Technology® for lightweight dexterity and cool comfort on the job.
  • Coreless DX+ Technology® achieves higher levels of cut protection without the discomfort that fiberglass or steel can cause
  • DX+ Technology® feels cool to the touch which keeps the wearer cool and comfortable while working
  • Polyurethane palm coating maintains dexterity while delivering excellent grip
  • Color-coded overcast acts as a visual aid for sizing and reduces fraying
  • Machine washable for longer service life
  • ANSI Level A6 cut resistance, ANSI Level 6 abrasion resistance, and ANSI Level 4 puncture resistance
DX+ Technology® uses strength-enhancing microparticles to deliver lightweight coreless cut protection with cooler wear and less skin irritation compared to fiberglass and steel core options. This leads to a 30% lighter glove compared to similar HPPE materials. Performance-wise, the Magid® D-ROC® DXPG62 resists cuts, abrasion, and punctures for optimal safety and is the only coreless glove that achieves ANSI Cut Level A6 resistance. The lightweight polyurethane palm coating maintains dexterity while delivering excellent grip.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty