Would you like to log in again?
Magid® ROC® GP210 Nylon Polyurethane Palm Coated Work Gloves
Supplier: Magid
Magid® ROC®
GP2108 GP2106 GP2109 GP2107 GP21011 GP21010
76635-780DZ 56.4 USD InStock
76635-780 76635-790 76635-784 76635-782 76635-788 76635-786
Magid® ROC® GP210 Nylon Polyurethane Palm Coated Work Gloves
Gloves
The GP210 is Magid's first 21-guage glove.
  • Polyurethane palm coating maintains dexterity while delivering excellent grip
  • Nylon shell adds stretch and breathability
  • Ultra-lightweight, form-fitting 21-gauge construction delivers exceptional comfort and the dexterity of bare hands
  • Color-coded overcast acts as a visual aid for sizing and helps prevent fraying
  • Machine washable for a longer service life
  • ANSI Level 1 Abrasion Resistance and ANSI Level 2 Puncture Resistance
This style finally delivers maximum comfort for those in need of a general-purpose glove. The GP210 provides a great grip with a polyurethane coating, and was designed with dexterity in mind for almost any general-purpose application.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty