Would you like to log in again?
Magid® T-REX® Flex Series® Knit Impact Gloves with NitriX Grip Palm, Cut Level A9
Supplier: Magid
Magid® T-REX® Flex Series®
TRX795XL TRX795XXL TRX795XXXL TRX795L TRX795M TRX795XXXXL
76635-838PR 36.3 USD InStock
76635-838 76635-842 76635-840 76635-836 76635-834 76635-844
Magid® T-REX® Flex Series® Knit Impact Gloves with NitriX Grip Palm, Cut Level A9
Gloves
The Magid® T-REX® Flex Series® TRX795 NitriX® Grip Technology Palm Impact Glove delivers high quality A9 cut resistance and Impact Level 1 protection at a great value.
  • 13-gauge Hyperon® shell delivers breathability, comfort, and cut protection
  • Nitrile reinforced thumb saddle enhances durability at a critical wear point
  • Elastic knit wrist cuff ensures a secure fit
  • Machine washable for longer service life
  • ANSI Level A9 Cut Resistance, ANSI Level 5 Abrasion Resistance and ANSI Level 4 Puncture Resistance
  • ANSI/ISEA 138 Impact Level 1 (Level 1 protection on knuckles and Level 1 protection on fingers)
NitriX® Grip Technology palm coating delivers rock-solid grip in oily applications. M-Force Defense System® back-of-hand protection extends all the way to the finger-tip and between thumb and index finger to deflect and absorb impacts.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty