Would you like to log in again?
HyClone™ Hanks' Balanced Salt Solutions, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyClone™
SH30268.LS SH30031.FS SH30588.02 SH30016.02 SH30588.01 SH30268.LS SH30015.02 SH30031.02 SH30031.03 SH30588.02 SH30031.03 SH30030.02 SH30031.01 SH30015.02 SH30015.03 SH30031.FS SH30015.03 SH30016.02 SH30031.01 SH30268.01 SH30268.02 SH30030.02 SH30268.02 SH30031.02 SH30268.02 SH30268.01 SH30588.01
76635-848EA 22.41 USD InStock
76635-848 CA82013-226 82024-308 CA16777-119 CA76635-852 CA16750-130 CA95053-196 CA16750-092 16777-153 CA18777-008 CA82020-069 16777-119 16777-159 82013-226 16777-114 CA76635-848 95053-196 76635-852 CA82024-308 16750-092 CA16777-153 82020-069 18777-008 82020-068 16750-130 CA16777-159 CA16777-114
HyClone™ Hanks' Balanced Salt Solutions, HyClone products (Cytiva)
Buffers Cell Culture Buffers
Balanced salt solutions for cell culture applications.
  • All formulations incorporate a rigorous specification and approval process
  • Powder formats rehydrate to specified quantity
  • Solutions: 0.1 µm sterile filtered
Buffer cell culture growth with Cytiva HyClone Hank's 1X Balanced Salt Solutions, available in lot sizes that facilitate scale-up and manufacturing.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty