Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HyClone™ CDM4NS0 Hybridoma and Myeloma Media, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH30578.01 SH30578.01
76635-874EA 482.48 USD InStock
CA76635-874 76635-874
HyClone™ CDM4NS0 Hybridoma and Myeloma Media, HyClone products (Cytiva)
Cell Culture Media
HyClone™ CDM4NS0 media was developed through metabolic pathway design approach to increase process yields in the manufacture of monoclonal antibodies (MAbs) for therapeutic use using a variety of cholesterol auxotrophic NS0 cell clones.
  • Chemically-defined, animal derived component free (ADCF) formulation
  • Complete, ready to use formulation
  • Allows for direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies
  • Component traceability
  • Manufactured according to cGMP guidelines
HyClone™ CDM4NS0 media require no cholesterol or GS supplementation, as the media have sufficient amounts to support NS0 cell culture already in the formulation. CDM4NS0 media have been successfully tested in a variety of culture systems, including T-flasks, shaker flasks, and bioreactors using batch and fed-batch strategies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty