Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HyClone™ PF-CHO LS Cell Culture Media, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH30334.01 SH30335.01 SH30359.02 SH30333.02 SH30335.01 SH30333.02 SH30333.01 SH30333.01 SH30359.02 SH30334.01
76635-872EA 217.75 USD InStock
76635-872 CA76462-522 CA16777-546 76462-524 76635-886 CA76635-872 16777-546 76462-522 CA76462-524 CA76635-886
HyClone™ PF-CHO LS Cell Culture Media, HyClone products (Cytiva)
Cell Culture Media
HyClone™ PF-CHO LS media is designed to support the DHFR selection/amplification system. It has been successfully tested in a variety of cell culture systems, including T-flasks, spinner flasks, and bioreactors.
  • Designed to support the DHFR selection/amplification system
  • Successfully tested in a variety of cell culture systems, including T-flasks, spinner flasks and bioreactors
  • Enable growth of many CHO cells with minimal adaptation
  • PF-CHO LS liquid medium contains 4 mM L-glutamine and 0,1% poloxamer 188
  • PF-CHO MPS powder medium does not contain L-glutamine or poloxamer 188
HyClone™ PF-CHO LS media is ideally suited for the cost-effective manufacture of recombinant proteins for academic and industrial applications, genomics and proteomics, in vitro diagnostics, drug target screening and validation, and manufacturing of pre-clinical lots.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty