Would you like to log in again?
ShuttlePouch, 95 kPa
Supplier: Therapak
CSTGSHU-02-0395KPA CSTGSHU-02-3-2 CSTGSHU-02-0695KPA
76635-920CS 1216.65 USD InStock
76635-920 76635-924 76635-922
ShuttlePouch, 95 kPa
Bags Sample Bags
Therapak’s 95 kPa transport pouches and bags are ideal to use as a secondary leakproof packaging in accordance with the Department of Transportation (DOT) and the International Air Transport Association (IATA) regulations for shipping Biological Substances.
  • Meets DOT and IATA requirements
  • Integrated absorbent sheet
  • Quick tear opening feature
DOT and IATA shipping regulations require that specimens classified as UN3373 Biological Substance, Category B are transported in either a primary receptacle or secondary packaging that has passed the 95 kPa pressure differential test as a part of the triple packaging regulations.
Therapak offers the innovative, award-winning ShuttlePouch for use with kits and packaging that require compliance to IATA Packing Instructions 650 for specimens classified as UN3373 Biological Substance, Category B.
The space-saving ShuttlePouch offers a vast reduction in plastic usage. It is easy to use and has a preinserted absorbent. It is ideal for shipping specimens back to the lab in the original kit packaging or alternate shippers. When ready to use, simply insert the specimen tube, follow the closing instructions printed on the pouch, and complete the kit packing instructions for secure and compliant transport. The pouch features an easy to open tear area to quickly access the specimen tube and facilitate processing when the samples arrive at the lab.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty