Would you like to log in again?
Evacuable KBr Pellet Dies, Firebird Optics
Supplier: Firebird Optics
KB-3 KBX-35 KB-10 KBX-32 KB-6 KBX-20 KB-16 KB-8 KBX-25 KBX-45 KB-13 KB-5 KBX-40 KB-7
76636-392EA 2981.06 USD InStock
76636-392 76636-350 76636-382 76636-352 76636-384 76636-340 76636-354 76636-386 76636-390 76636-356 76636-388 76636-344 76636-338 76636-348
Evacuable KBr Pellet Dies, Firebird Optics
Presses
Dies suitable for producing discs of 3, 5, 6, 7, 8, 10, 13, 16, 20, 25, 32, 35 and 40 mm diameter as available as standard within the size range for the KB series. Non-standard sizes are available on request.
  • All dies in the KB series are constructed entirely of stainless tool steel, and parts are precision machined
  • Pellets are optically flat and mirror polished
  • Tungsten carbide pellets and component parts can be supplied to special order
  • Dies are supplied with black anodised sleeves
KBr Disc Technique: This widely practiced method consists of grinding and mixing a sample with dry KBr or KCl powder, which is then compressed within an evacuated die to produce a transparent disc, since KBr and KCl are transparent from the ultra-violet well into the infrared. The method may be used for investigation in the ultra-violet and the visible as well as the infrared region.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty