Would you like to log in again?
Water-Jacketed Evacuable KBr Pellet Dies, Firebird Optics
Supplier: Firebird Optics
KB-WJ10 KBX-WJ20 KBX-WJ32 KB-WJ6 KBX-WJ35 KB-WJ8 KBX-WJ40
76636-380EA 1652.42 USD InStock
76636-380 76636-372 76636-394 76636-398 76636-378 76636-400 76636-402
Water-Jacketed Evacuable KBr Pellet Dies, Firebird Optics
Presses
Water-jacketed dies suitable for producing discs of 3, 5, 6, 7, 8, 10, 13, 16, 20, 25, 32, 35 and 40 mm diameter as available as standard within the size range for the KB series. Non-standard sizes are available on request.
  • All dies in the KB series are constructed entirely of stainless tool steel, and parts are precision machined
  • Pellets are optically flat and mirror polished
  • Tungsten carbide pellets and component parts can be supplied to special order
  • Dies are supplied with black anodised sleeves
Allows heating of solid samples while under pressure, when electrical appliances cannot be used. Boiling water is passed through flow through tube and circulates heating the die body to between 80..100 ºC. Water jacket can also be used to cool sample under pressure.
KBr Disc Technique: This widely practiced method consists of grinding and mixing a sample with dry KBr or KCl powder, which is then compressed within an evacuated die to produce a transparent disc, since KBr and KCl are transparent from the ultra-violet well into the infrared. The method may be used for investigation in the ultra-violet and the visible as well as the infrared region.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty