Would you like to log in again?
Coriolis® Air Sampler Accessories, Bertin Instruments
Supplier: Bertin
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Coriolis®
S001108-CORM0-A.0 S001112-CORM0-A.0 S001109-CORM0-A.0 S001110-CORM0-A.0 S001113-CORM0-A.0 S002345-CORC0-A.0 S001111-CORM0-A.0
76636-440PK 573.18 USD InStock
76636-440 76636-450 76636-430 76636-432 76636-434 76636-436 76636-438
Coriolis® Air Sampler Accessories, Bertin Instruments
Samplers Air Samplers
Coriolis® air sampler consumables are part of the innovative dry cyclonic technology, which aspirates airborne particles and centrifuges them in a cone.
  • Single use consumables
  • Ensure optimal collection efficiency for Coriolis® air samplers
The separation of the airborne particles from the air flow is due to the air flow rate, the air intake geometry, the design of the cones, and the collection liquid (surfactant in low concentration). The collected particles or microorganisms can then be recovered by rinsing the consumables with an appropriate buffer solution, so the sample is compatible with multiple downstream analysis options (NGS, qPCR, culture).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty