Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® ULT Freezers Eco Premium 2" Cryobox Storage Bundles, 208 V

Supplier: VWR
VWR®
9020-8468 9020-7468 9020-8467 9020-7469 9020-7467 9020-8469
76636-490EA 14239.94 USD InStock
76636-490 76636-492 76636-494 76636-488 76936-178 76936-176
VWR® ULT Freezers Eco Premium 2" Cryobox Storage Bundles, 208 V
Freezers Ultra Low Temperature Freezers
The VWR® ULT Freezers Eco Premium are designed for safe and reliable storage of sensitive samples. They feature ergonomic control and low energy consumption.

  • Maximum 2" box capacity: 352/528
  • Energy star certified
  • Temperatures between −86 to −50 °C
  • Long warm-up time (warm up time from −86 to −50 °C: 360 min)
  • Highly efficient VIP insulation and long-lasting rust-proof sample chamber
  • Green refrigerants
  • USB and Ethernet interface for data transfer

The VWR® ULT Freezers Eco Premium have a stainless steel inner chamber, flexible rack configuration and are further characterized by low energy consumption and electricity costs. They have individually adaptable chamber compartments and samples remain frozen for >30 hours during power outage or system failure. The climate-friendly refrigerants used help to protect the environment.

Warranty: VWR® Five-year limited parts and labor warranty; VWR® Five-year warranty on compressor parts.

Certifications: UL, cUL, CE marked, UKCA.

Ordering information: A variety of accessories are available, these must be ordered separately.

Delivery information: VWR® ULT Freezers are supplied with racks for 2" standard boxes for full freezer load.

Packaging: VWR® ULT Freezer plus a package of racks for 2" standard boxes.

Caution: Note these are 208 V units

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty