Would you like to log in again?
Cryo Racks, Open Bottom, Barcoded, Azenta Life Sciences
Supplier: Azenta Life Sciences
66-1803 66-1800 66-1802 66-0196-03 66-1801 66-0196-02
77320-730CS 211.85 USD InStock
77320-730 76636-576 76636-982 76636-720 76636-624 76637-072
Cryo Racks, Open Bottom, Barcoded, Azenta Life Sciences
Racks Cryogenic Vial Racks
Open bottom for 2D-code decoding on camera-based readers.
Direct laser etched 1D linear barcode and human-readable number on rack side and 2D rack ID on base of rack.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty