Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dual-Coded Sample Storage Tubes, 0.26 ml, 96-format, External Thread, Azenta Life Sciences

Supplier: Azenta Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

68-0303-11 68-0303-01 68-0303-10 68-0303-14 68-0303-13 68-0303-00
76636-590CS 1114.77 USD InStock
76636-590 76636-584 76636-650 76636-796 76636-656 76636-956
Dual-Coded Sample Storage Tubes, 0.26 ml, 96-format, External Thread, Azenta Life Sciences
Vials
Azenta Life Sciences dual-coded tubes feature a 2D-code and Human Readable Number (HRN) on the tube base, allowing compatibility with low throughput manual workflows, semi-automated workflows or fully automated workflows on integrated platforms.

  • Secure sample storage and tracking
  • Permanently laser etched, 2D and human readable codes
  • 100% Quality Control, each tube is tested to ensure both readability and uniqueness
  • Manufactured using an advanced process which results in a one-piece jacket tube from high-quality virgin polypropylene
  • Suitable for cryogenic storage as low as –196 °C (not for use in liquid phase nitrogen)
  • Secure sample storage and tracking
  • 2D-codes readable without removing tubes from racks

Designed and developed with broad compatibility in mind, the tubes perform without compromise in conjunction with automated barcode reading, capping and sample management systems from Azenta Life Sciences and all other industry-recognized manufacturers. Provides a lifelong and secure chain of custody for samples in biobanks, compound libraries and a broad range of biological and chemical stores, including cryogenic storage.

Each tube carries a unique and permanent high-contrast 2D code tube identifier on the base of the tube readable in frost or condensation conditions, or when damaged. Manufactured from high-quality virgin polypropylene.

Ordering information: Available bulk uncapped or capped.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty