Would you like to log in again?
Tri-Coded Sample Storage Tubes, 1.9 ml, 48-format, External Thread, Azenta Life Sciences
Supplier: Azenta Life Sciences
65-7643-EB 65-7643 65-7642 65-7640 65-7641 65-7643-GS
76636-826CS 638.45 USD InStock
76636-826 76636-552 76636-794 76636-564 76637-064 76637-084
Tri-Coded Sample Storage Tubes, 1.9 ml, 48-format, External Thread, Azenta Life Sciences
Vials
Azenta Life Sciences tri-coded tubes offer unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities.
  • Secure sample storage and tracking
  • Permanently laser etched, 2D-code on base, 1D (linear barcode) and Human-Readable Number on the side
  • Tri-coded offers unequalled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities
  • 100% Quality Control, each tube is tested to ensure both readability and uniqueness
  • Manufactured using an advanced process which results in a one-piece jacket tube from high-quality virgin polypropylene
  • Suitable for cryogenic storage as low as –196 °C (not for use in liquid phase nitrogen)
  • 2D-codes readable without removing tubes from racks
  • Suitable for sealing with either screw caps or TPE septum caps
Offers unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities. Designed and developed with broad compatibility in mind, the tubes perform without compromise in conjunction with automated barcode reading, capping and sample management systems from Azenta Life Sciences and all other industry-recognized manufacturers.
Provides a lifelong and secure chain of custody for samples in biobanks, compound libraries and a broad range of biological and chemical stores, including cryogenic storage.
Developed to exceed the demands of sample security, management and tracking in modern high density storage applications.
Ordering information: Available bulk uncapped or capped.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty