Would you like to log in again?
Sample Storage Tube Caps, Screw Caps
Supplier: Azenta Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
68-53100-Z11N 66-63100-Y6 68-53100-Z4N 66-63100-Y13 66-9401-S 65-7575 68-53100-Z1N 66-9401 66-63100-Y1 66-63100-Y5 68-53100-Z2N 66-63100-Y2 68-53100-Z10N 65-7573 66-63100-Y12 65-9373 65-7572 68-53100-Z8N 66-63100-Y10 66-63100-Y11 68-53100-Z5N 65-7574 65-7576 68-53100-Z3N 66-63100-Y3 66-63100-Y4 66-9402 68-53100-Z12N 67-0202-50 67-0203-51 66-63100-Y8 68-53100-Z13N 68-53100-Z6N
76636-690CS 261.15 USD InStock
76636-690 76637-114 76636-992 76636-606 76636-906 76636-620 76637-122 76636-644 76636-600 76636-966 76637-106 76637-002 76636-914 76636-936 76636-638 76636-718 76636-972 76636-652 76637-110 76637-054 76636-754 76636-996 76636-896 76636-932 76636-712 76636-976 76636-854 76636-736 76636-934 76636-714 76637-094 76636-636 76636-812
Sample Storage Tube Caps, Screw Caps
Closures Test/Sample Tube Closures
Tube screw caps are manufactured from high-quality virgin polypropylene and are designed for optimal seal quality and sample security.
  • High chemical resistance
  • Broad operating temperature range –196 to +121 °C, do not use in liquid phase nitrogen
  • Automation friendly, available in Azenta cap carrier for use with automated capping and de-capping systems
  • Autoclavable
  • Available in up to 10 different colors to aid sample identification
Developed to exceed the demands of sample security, management and tracking in modern high-density storage applications.
Compatible with all Azenta 96-format sample storage tubes with a screw top, caps are available for use with either external or internal thread Azenta screw top tubes.
Ordering information: These tubes are supplied in bags of 960 caps.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty