Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sample Storage Tube Caps, TPE Septum Caps

Supplier: Azenta Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

65-73001-S 65-73001-10 65-75000 65-76000 65-73003 65-73001 65-74000 65-73004-10 65-73003-10 65-73002 65-73004 65-73002-10 65-73000-10 65-73000
76636-596CS 304.56 USD InStock
76636-596 76636-738 76636-968 76637-012 76636-554 76636-730 76636-894 76636-732 76636-930 76636-742 76637-130 76636-844 76636-766 76636-612
Sample Storage Tube Caps, TPE Septum Caps
Closures Test/Sample Tube Closures
TPE septum caps are piercable, disposable, thermo plastic elastomer (TPE) caps designed for optimal seal quality.

  • Broad operating temperature range, suitable for use −80 to +110 °C
  • Ideal solution for –20 °C storage with occasional access
  • Available in a choice of five colors to aid sample identification
  • Natural color only recommended for automatic systems
  • Supplied in 96-format back mats to facilitate automation
  • Septum cap compatibilty

Developed to meet the needs of sample security, management and tracking in modern high-density storage applications. Compatible with 96-format tubes, TPE caps are available for use with internal thread tubes and are manufactured from high-quality TPE, supplied as 96-cap mats or in bulk.

Suitable for automatic and semi-automatic cappers and de-capper.
Suitable for 96-format Tri-coded Sample Storage Tubes, internal thread 0.65, 0.9 ml.
Suitable for 96-format Dual-coded Tubes internal thread 0.3, 0.7 and 0.9 ml.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty