Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Non-Coded Sample Storage Tubes, 0.3 ml, 96-format, Internal Thread, Azenta Life Sciences

Supplier: Azenta Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

66-32141 66-32041-Y6 66-32041-Y6-L 66-32040 66-32041 66-32040-Y6
76637-080CS 674 USD InStock
76637-080 76637-026 76637-078 76636-874 76636-668 76636-878
Non-Coded Sample Storage Tubes, 0.3 ml, 96-format, Internal Thread, Azenta Life Sciences
Vials
The same storage tubes are also available in non-coded format for independent customization.

  • Manufactured from high-quality virgin polypropylene
  • Suitable for cryogenic storage as low as –196 °C (not for use in liquid phase nitrogen)
  • Secure sample storage and tracking

Ordering information: Available bulk uncapped or capped.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty