Would you like to log in again?
Grippaz™ Extended Use Ambidextrous Nitrile Gloves, Protective Industrial Products
Supplier: Protective Industrial Products
Grippaz™
67-246/M 67-306/XL 67-246/S 67-246/XXL 67-307/L 67-306/S 67-246/XL 67-307/M 67-307/2XL 67-307/3XL 67-307/XL 67-307/S 67-306/M 67-246/L 67-306/L
76637-168CS 153.8 USD InStock
76637-168 76637-178 76637-180 76637-190 76637-166 76637-188 76637-186 76637-164 76637-174 76637-184 76637-162 76637-172 76637-182 76637-170 76637-192
Grippaz™ Extended Use Ambidextrous Nitrile Gloves, Protective Industrial Products
Gloves
Grippaz™ Technology features a fish scale pattern that provides unparalleled grip performance. Ergonomically designed to provide traction grip on the outside and on the inside Tough stretch formula nitrile fits more snugly than traditional gloves. These gloves offer maximum comfort, multiple use durability, and dexterity. Ideal for handling oily/wet parts, paint spraying, automotive service and repair, and electroplating objects.
  • Ambidextrous nitrile gloves
  • Powder-free and silicone-free
  • Patented fish scale grip
  • FDA 21 CFR compliant
Patented design on the inside and outside of the glove provides extra traction with oily or wet parts. These 6 mil or 7 mil proprietary nitrile blended gloves feature a patented fish scale pattern that is more tear-resistant and puncture-resistant than conventional single-use disposable gloves. This design allows prolonged use and reduced waste.
ESD Safe, Surface Resistance per ASTM D257, Static dissipative range at 10⁸ ohms, 12% RH, 48 h @100 V. Resistant to many chemicals commonly used in Industrial and OEM applications including oils, grease, and cleaning solvents. The proprietary nitrile blend offers enhanced flexibility, comfort, and mobility to reduce hand fatigue. Touchscreen compatible to allow the user to operate a touch screen phone or device without removing gloves.
Black finish effectively hides dirt. Yellow finish is synonymous with chemical handling. Orange finish provides enhanced eye-to-hand visual acuity.
Certifications: Complies with FDA food handling requirements 21 CFR, Part 177.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty