Would you like to log in again?
CR2700 Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, Brady Worldwide
Supplier: Brady
176516 176515
76637-592EA 1932.93 USD InStock
76637-592 76637-538
CR2700 Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, Brady Worldwide
Barcode Systems
Easily read wide and small barcodes with the ultra-fast CR2700 Handled barcode scanner. This aggressive, omnidirectional scanner eliminates repositioning and time consuming re-scans of barcodes.
  • Uses B27 batteries with built-in power gauge
  • IP65 seal rating against dust and liquid ingress
  • CodeShield® housing for disinfectant and chemical wipe down
  • Unequalled performance reading on curved and shiny surfaces
  • USB, Bluetooth®5 Low Energy (for enhanced data security, extended battery life, and less downtime)
Customizable visual, audible and haptic feedback with multiple programmable buttons. Trace any troubles with trackable service dates and battery statistics to help IT deploy and manage their scanner fleet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty