Would you like to log in again?
CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code, Brady Worldwide
Supplier: Brady
176522
76637-544EA 331.36 USD InStock
76637-544
CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code, Brady Worldwide
Barcode Systems
Improve data collection operations with this swift and accurate barcode reader. The CR950 thrives in fast-paced environments, offering increased efficiency with its intuitive targeting device. This kit features a Stand.
  • Swift and accurate barcode reader
  • Kit includes a stand
  • Manual or automatic triggering
  • Provides easier scanning with a wide area image sensor
  • Features two programmable indicators: LED and tone
  • 5-year limited warranty
The CR950 thrives in fast-paced environments, offering increased efficiency with its intuitive targeting device. Get high-speed omnidirectional reading of 1D and 2D barcodes and glare reduction technology for reading barcodes on shiny surfaces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty