Would you like to log in again?
Lockout Station with Nylon Safety Lockout Padlocks, Brady Worldwide
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
153684 153683 153682 153681
76637-558KT 202.48 USD InStock
76637-558 76637-532 76637-552 76637-540
Lockout Station with Nylon Safety Lockout Padlocks, Brady Worldwide
Security Locks
Safely power down faster when powering up lockout tagout readiness with this fully outfitted, easily installed, organized and accessible lockout station.
  • OSHA compliance driver
  • Built to last
  • Ultimate accessibility
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 control of hazardous energy. All components meet Brady’s highest standards for quality, durability and reliability. Place locks, tags and devices at the point of need to help authorized workers maintain safety and compliance. Comes with predrilled holes for easy wall mounting, and a carrying handle when it is needed on the go.
Ordering information: Lockout station 153684 includes: (12) nylon safety lockout padlocks, keyed alike, (1) lock and device station, (2) 1" lockout hasps, (2) 1.5" lockout hasp, (20) lockout tags, and (20) zip ties.
Lockout station 153683 includes: (12) nylon safety lockout padlocks, keyed different, (1) lock and device station, (2) 1" lockout hasps, (2) 1.5" lockout hasps, (20) lockout tags, and (20) zip ties.
Lockout station 153682 includes: (6) nylon safety lockout padlocks, keyed alike, (1) lock and device station, (2) 1" lockout hasps, (10) lockout tags, and (10) zip ties.
Lockout station 153681 includes: (6) nylon safety lockout padlocks, keyed different, (1) lock and device station, (2) 1" lockout hasps, (10) lockout tag, and (10) zip ties.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty