Would you like to log in again?
Electrical and Valve Lockout Tagout Kit with Nylon Safety Lockout Padlocks in Duffel Bag, Brady Worldwide
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
153672
76637-598KT 941.61 USD InStock
76637-598
Electrical and Valve Lockout Tagout Kit with Nylon Safety Lockout Padlocks in Duffel Bag, Brady Worldwide
Security Locks
Safely power down faster when powering up lockout tagout readiness with this convenient, easily accessible and portable lockout kit.
  • OSHA compliance driver
  • Built to last
  • Extremely portable
  • Great for contractors
  • Sores easily in vehicles to travel between jobsites
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 Control of Hazardous Energy. All components meet Brady’s highest standards for quality, durability and reliability.
Duffel keeps padlocks and devices at the ready and goes virtually anywhere. Organize, transport, and deploy everything needed to lock out breakers, plugs, and valves in a multitude of types and sizes with this robust kit. Each lock and key are unique, allowing authorized users to have their own individual locks and comply with OSHA’s 'one person, one lock, one key' guidance.
Ordering information: Lockout Kit includes: (3) Nylon Safety Lockout Padlocks, (1) Lockout Duffel, (2) Lockout Pouches, (2) Lockout Hasps, (1) Cable Lockout with 8' Metal Cable, (1)Universal Valve Lockout with Blocking Arm, (1) Gate Valve Lockout 1 - 2.5", (1) Gate Valve Lockout 2.5- 5", (1) Gate Valve Lockout 5 - 6.5", (1) Ball Valve Lockout, (2) Clamp-on Circuit Breaker Lockout 120/277 V, (1) Clamp-on Circuit Breaker Lockout 480/600V, (2) Universal Multipole Breaker Lockout, (1) 3-in-1 Plug Lockout, (1) Wall Switch Lockout, (10) Lockout Tags, and (10) Zip Ties.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty