Would you like to log in again?
CR2700 Handheld Wireless Palm Barcode Scanner: Bluetooth, 1D, 2D, QR Code, Brady Worldwide
Supplier: Brady
176518 176517
76637-610EA 1932.93 USD InStock
76637-610 76637-554
CR2700 Handheld Wireless Palm Barcode Scanner: Bluetooth, 1D, 2D, QR Code, Brady Worldwide
Barcode Systems
Read wide and small barcodes easier with this ultra-fast barcode reader. Get unbeatable performance from this aggressive, omnidirectional scanner that reads even damaged barcodes.
  • Uses B27 batteries with built-in power gauge
  • IP65 seal rating against dust and liquid ingress
  • CodeShield® housing for disinfectant and chemical wipedown
  • Unequaled performance reading on curved and shiny surfaces
  • USB, Bluetooth®5 Low Energy (for enhanced data security, extended battery life, and less downtime)
Customizable visual, audible and haptic feedback with multiple programmable buttons. Trace any troubles with trackable service dates and battery statistics to help IT deploy and manage their scanner fleet.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty