Would you like to log in again?
Nylon Safety Lockout Padlock and Tag Kit, Brady Worldwide
Supplier: Brady
153673
76637-630KT 14.94 USD InStock
76637-630
Nylon Safety Lockout Padlock and Tag Kit, Brady Worldwide
Security Locks
Stock up on this slim kit with its uniquely keyed padlock and reusable tag that helps keep any worker ready and compliant with OSHA’s ‘one person, one lock, one key’ guidance.
  • OSHA compliance driver
  • Built to last
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 control of hazardous energy. All components meet Brady’s highest standards for quality, durability and reliability.
Reusable heavy-duty polyester tag is protected by write-on laminate that wipes clean and withstands water, grease, and extreme temperatures. Key-retaining nylon padlock helps prevent lost keys as well as electricity traveling from shackle to key.
Ordering information: Lockout kit includes: (1) nylon safety lockout padlock, (1) lockout tag, and (1) zip tie.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty