Would you like to log in again?
Lockout Tagout Kit with Nylon Safety Padlock in Pouch, Brady Worldwide
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
153668
76637-640KT 111.35 USD InStock
76637-640
Lockout Tagout Kit with Nylon Safety Padlock in Pouch, Brady Worldwide
Security Locks
Safely power down faster when you power up your lockout tagout readiness with one convenient, easily accessible and portable kit.
  • OSHA compliance driver
  • Built to last
  • Extremely portable
  • Great for contractors
  • Stores easily in vehicles to travel between jobsites
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 control of hazardous energy. All components meet Brady’s highest standards for quality, durability and reliability.
Belt-attachable pouch keeps tools at the ready and goes anywhere.
Ordering information: Lockout kit includes: (1) nylon safety lockout padlock, (1) lockout pouch, (2) 1" lockout hasps, (5) lockout tags, and (5) zip ties.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty