Would you like to log in again?
Electrical Lockout Tagout Kit with Nylon Safety Lockout Padlocks in Toolbox, Brady Worldwide
Supplier: Brady
153670
76637-644KT 137.94 USD InStock
76637-644
Electrical Lockout Tagout Kit with Nylon Safety Lockout Padlocks in Toolbox, Brady Worldwide
Security Locks
Safely power down faster when you power up your lockout tagout readiness with one convenient, easily accessible and portable kit.
  • OSHA compliance driver
  • Built to last
  • Extremely portable
  • Great for contractors
  • Stores easily in vehicles to travel between jobsites
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 control of hazardous energy. All components meet Brady’s highest standards for quality, durability and reliability.
Toolbox keeps padlocks and devices at the ready and goes anywhere. All components are specially designed for electrical lockout scenarios, including non-conductive nylon-body padlocks. Contains two each of customer-favorite circuit breaker lockouts covering 120/277 V and 480/600 V in single and multi-pole configurations. Each lock and key are unique, allowing authorized users to have their own individual locks and comply with OSHA’s 'one person, one lock, one key' guidance.
Ordering information: Lockout kit includes: (3) nylon safety lockout padlocks, (1) lockout toolbox, (2) 1" lockout hasps, (2) clamp-on circuit breaker lockouts 120/277 V, (1) clamp-on circuit breaker lockout 480/600 V, (2) universal multipole breaker lockouts, (1) 3-in-1 Plug lockout, (1) wall switch lockout, (10) lockout tags, and (10) zip ties.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty