Would you like to log in again?
Electrical Lockout Station with Safety Lockout Padlocks, Brady Worldwide
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
153686 153685
76637-646KT 421.09 USD InStock
76637-646 76637-546
Electrical Lockout Station with Safety Lockout Padlocks, Brady Worldwide
Security Locks
Safely power down faster when powering up lockout tagout readiness with this fully outfitted, easily installed, organized and accessible electrical lockout station.
  • OSHA compliance driver
  • Built to last
  • Ultimate accessibility
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 control of hazardous energy. All components meet Brady’s highest standards for quality, durability and reliability. Place locks, tags and devices at the point of need to help authorized workers maintain safety and compliance. Comes with predrilled holes for easy wall mounting, and a carrying handle when it is needed on the go.
Ordering information: Lockout station 153686 includes: (6) nylon safety lockout padlocks, keyed alike, (1) lock and device station, (2) 1" lockout hasps, (2) 1.5" lockout hasps, (1) cable lockout with 8' metal cable, (2) clamp-on circuit breaker lockout 120/277 V, (1) clamp-on circuit breaker lockout 480/600 V, (2) universal multipole breaker lockouts, (1) 3-in-1 plug lockout, (2) wall switch lockouts, (20) lockout tags, and (20) zip ties.
Lockout station 153685 includes: (6) nylon safety lockout padlocks, keyed different, (1) lock and device station, (2) 1" lockout hasps, (2) 1.5" lockout hasps, (1) cable lockout with 8' metal cable, (2) clamp-on circuit breaker lockouts 120/277 V, (1) clamp-on circuit breaker lockout 480/600 V, (2) universal multipole breaker lockouts, (1) 3-in-1 plug lockout, (2) wall switch lockouts, (20) lockout tags, and (20) zip ties.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty