Would you like to log in again?
CR1500 Handheld Wired Barcode Scanner and Barcode Scanner with Stand; 1D, 2D, QR Code, Brady Worldwide
Supplier: Brady
176511 176510
76637-656EA 538.48 USD InStock
76637-656 76637-576
CR1500 Handheld Wired Barcode Scanner and Barcode Scanner with Stand; 1D, 2D, QR Code, Brady Worldwide
Barcode Systems
Save valuable time with a fast and reliable scanning solution. This omnidirectional barcode reader eliminates repositioning and time consuming re-scans of 1D and 2D barcodes that have seen challenging conditions.
  • Compact, lightweight and rugged to survive the toughest industrial environments
  • Fast and easy scanning with a high-performance scan engine and intuitive targeting
  • Read both high-density and wide-field barcodes with a single device via dual-field optics
Smart scanning in a small package, the CR1500 handheld wired barcode scanner is one of the smallest barcode readers on the market. The PVC-free housing is IP 54 rated to protect against harsh chemical cleaning agents, dust, and water ingress.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty