Would you like to log in again?
Gamma Wipe® Polx® Nonwoven 100% Polyester Cleanroom Wipers, Berkshire
Supplier: Berkshire
Gamma Wipe® Polx®
GWPNWST1242
76637-684CS 375 USD InStock
76637-684
Gamma Wipe® Polx® Nonwoven 100% Polyester Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Gamma Wipe® Polx® Nonwoven is a 100% polyester hydroentangled nonwoven wiper recommended for ISO Class 5 and above environments.
  • Exceptional cleanliness
  • Gamma irradiated and sterile validated to a 10¯⁶ Sterility Assurance Level per AAMI guidelines
  • Chemically compatible with commonly used disinfectants and sporicides
  • Prevents abrasion with its extremely soft and pliable surface
  • Works well with IPA and other solvents
  • Knife cut
  • Lot to lot traceability
  • Autoclavable
Gamma Wipe® Polx® Nonwoven wipers are extremely soft and pliable, and ideal for use on all types of sensitive surfaces and an excellent choice for wrapping delicate parts for protection. Additionally, it's critically low fiber and particle generation and minimal non-volatile residues make Gamma Wipe® Polx® Nonwoven an exceptional choice for streak-free cleaning.
Certifications: ISO Class 5 to 8, Cleanroom Class 100 to 100,000, EU Grade A/B/C/D.
Ordering information: Certificate of Sterility with each case and available on the web by lot number 24/7. Details: Expiration Date, Radiation Dosage, Lot Information, Meets cGMP requirements for traceability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty