Would you like to log in again?
Heat Sealing Films, Clear, Easily Pierceable
Supplier: Azenta Life Sciences
4TI-0581 4TI-0580/S 4TI-0582 4TI-0580 4TI-0580/122 4TI-0582/S
76637-702EA 1217.37 USD InStock
76637-702 76637-888 76637-820 76637-720 76637-692 76637-790
Heat Sealing Films, Clear, Easily Pierceable
Seals Microplate Seals
The clear heat seal easily pierceable is an optically clear polyester backed film, forming a pierceable seal to polypropylene, polyethylene, polystyrene and cyclic olefin copolymer (COC) plates. For short term compound storage. Suitable for PCR, qPCR and optical applications.
  • Thin polyester heat sealing film
  • Easily pierceable with autosampler needles/ABI® 3730
  • Compatible with automation and for use on needle, capillary and tip based liquid handling systems
  • Moderate solvent resistance
  • Perforated roll for easy removal of sheets, for use with manual and semi-automated sealers
  • Temperature range: –80 to +80 °C
  • Non sterile
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. The optical clarity of this seal enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays.
The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.
Ordering information: Available in multiple roll formats compatible with specified automated heat sealers. Gamma treatment available upon request.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty